Cyclopentanecarbonitrile ≥98.0% (by GC)
Supplier: TCI America
|
Danger
|
C3167-25ML
C3167-5ML
TCC3167-25MLEA
517
USD
TCC3167-25ML
TCC3167-5ML
Cyclopentanecarbonitrile ≥98.0% (by GC)
Cyclopentanecarbonitrile
Formula:
C₆H₉N Boiling Pt: 168 °C Flash Pt: 56 °C |
MDL Number:
MFCD00013741 CAS Number: 4254-02-8 EINECS: 224-229-2 UN: 1992 ADR: 3,III |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...