Benzyl isovalerate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Benzyl 3-methylbutanoate

B6099-100G B6099-25G
TCB6099-100GEA 118 USD
TCB6099-100G TCB6099-25G
Benzyl isovalerate ≥98.0% (by GC)
Benzyl isovalerate
Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
Boiling Pt: 245 °C
MDL Number: MFCD00048348
CAS Number: 103-38-8
EINECS: 203-106-7

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR