3-Chloro-5-fluorophenylacetonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 3-Chloro-5-fluorophenylacetonitrile, 2-(3-Chloro-5-fluorophenyl)acetonitrile, 3-Chloro-5-fluorobenzyl cyanide

C3330-1G C3330-5G
TCC3330-1GEA 54 USD
TCC3330-1G TCC3330-5G
3-Chloro-5-fluorophenylacetonitrile ≥98.0% (by GC)
3-Chloro-5-fluorophenylacetonitrile
Formula: C₈H₅ClFN
MW: 169.59 g/mol
MDL Number: MFCD04038259
CAS Number: 493038-93-0
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR