Dibutyl carbonate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Carbonic acid dibutyl ester

C0040-25G C0040-5G
TCC0040-25GEA 191 USD
TCC0040-25G TCC0040-5G
Dibutyl carbonate ≥98.0% (by GC)
Dibutyl carbonate
Formula: C₉H₁₈O₃
MW: 174.24
Boiling Pt: 207 °C
MDL Number: MFCD00043938
CAS Number: 542-52-9
EINECS: 208-816-0
Merck Index: 14,01558

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR