2-Acetyl-2-thiazoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-(4,5-Dihydrothiazol-2-yl)ethanone

A3264-1G A3264-5G
TCA3264-1GEA 58 USD
TCA3264-1G TCA3264-5G
2-Acetyl-2-thiazoline ≥98.0% (by GC, titration analysis)
2-Acetyl-2-thiazoline
Formula: C₅H₇NOS
MW: 129.18 g/mol
Boiling Pt: 94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00209496
CAS Number: 29926-41-8

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR