Cyclohexene-1-carbonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 1-Cyclohexene-1-carbonitrile, Cyclohex-1-enecarbonitrile

C3191-1G C3191-5G
TCC3191-1GEA 54 USD
TCC3191-1G TCC3191-5G
Cyclohexene-1-carbonitrile ≥98.0% (by GC)
Cyclohexene-1-carbonitrile
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 74 °C
Flash Pt: 83 °C
MDL Number: MFCD00013772
CAS Number: 1855-63-6
EINECS: 217-454-2
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR