Dimethyl phenylphosphonite ≥98.0% (by GC)
Supplier: TCI America
|
Danger
|
D5014-1G
D5014-5G
TCD5014-1GEA
41
USD
TCD5014-1G
TCD5014-5G
Dimethyl phenylphosphonite ≥98.0% (by GC)
Dimethyl phenylphosphonite
Formula:
C₈H₁₁O₂P Boiling Pt: 102 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008352 CAS Number: 2946-61-4 EINECS: 220-960-6 UN: 1760 ADR: 8,II |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...