3-Bromo-4-formylpyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-Bromo-4-pyridinecarboxaldehyde, 3-Bromoisonicotinaldehyde, 3-Bromopyridine-4-carboxaldehyde

B2624-1G B2624-5G
TCB2624-1GEA 41 USD
TCB2624-1G TCB2624-5G
3-Bromo-4-formylpyridine ≥98.0% (by GC)
3-Bromo-4-formylpyridine
Formula: C₆H₄BrNO
MW: 186.01 g/mol
MDL Number: MFCD05864506
CAS Number: 70201-43-3

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 78.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR