2-Chloro-4-nitrobenzonitrile ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
C3122-1G
C3122-5G
TCC3122-1GEA
61
USD
TCC3122-1G
TCC3122-5G
2-Chloro-4-nitrobenzonitrile ≥98.0% (by GC)
2-Chloro-4-nitrobenzonitrile
Formula:
C₇H₃ClN₂O₂ |
MDL Number:
MFCD00052203 CAS Number: 28163-00-0 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 79.0 to 83.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...