3-Bromo-5-hydroxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B4740-1G B4740-5G
TCB4740-1GEA 50 USD
TCB4740-1G TCB4740-5G
3-Bromo-5-hydroxybenzoic acid ≥98.0% (by GC, titration analysis)
3-Bromo-5-hydroxybenzoic acid
Formula: C₇H₅BrO₃
MW: 217.02 g/mol
MDL Number: MFCD06797980
CAS Number: 140472-69-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 237.0 to 241.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR