2,6-Difluoro-3-nitrobenzonitrile ≥97.0% (by GC)

Supplier: TCI America
Danger

D5185-1G D5185-5G
TCD5185-1GEA 21 USD
TCD5185-1G TCD5185-5G
2,6-Difluoro-3-nitrobenzonitrile ≥97.0% (by GC)
2,6-Difluoro-3-nitrobenzonitrile
Formula: C₇H₂F₂N₂O₂
MDL Number: MFCD00042254
CAS Number: 143879-77-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 51.0 to 55.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR