4-Chloro-3-iodobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C3484-1G C3484-5G
TCC3484-1GEA 32 USD
TCC3484-1G TCC3484-5G
4-Chloro-3-iodobenzoic acid ≥98.0% (by GC, titration analysis)
4-Chloro-3-iodobenzoic acid
Formula: C₇H₄ClIO₂
MDL Number: MFCD00079732
CAS Number: 42860-04-8

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 216.0 to 220.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR