4-(Diethylamino)benzonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

D4852-25G D4852-5G
TCD4852-25GEA 138 USD
TCD4852-25G TCD4852-5G
4-(Diethylamino)benzonitrile ≥98.0% (by GC, titration analysis)
4-(Diethylamino)benzonitrile
Formula: C₁₁H₁₄N₂
MDL Number: MFCD00019754
CAS Number: 2873-90-7
EINECS: 220-712-7
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 66.0 to 70.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR