2-Chlorophenethyl bromide ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-(2-Bromoethyl)-2-chlorobenzene, 2-Chlorophenethyl Bromide, 2-(2-Chlorophenyl)ethyl bromide, 2-Chlorophenethyl bromide

B5833-1G B5833-5G
TCB5833-1GEA 34 USD
TCB5833-1G TCB5833-5G
2-Chlorophenethyl bromide ≥98.0% (by GC)
2-Chlorophenethyl bromide
Formula: C₈H₈BrCl
MW: 219.51 g/mol
Boiling Pt: 112 °C
MDL Number: MFCD04117464
CAS Number: 16793-91-2

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR