2-Chloro-4-pyridineboronic acid pinacol ester ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Chloropyridine-4-boronic acid pinacol ester, 2-(2-Chloro-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Chloropyridine-4-boronic acid Pinacol ester

C3529-1G C3529-5G
TCC3529-1GEA 66 USD
TCC3529-1G TCC3529-5G
2-Chloro-4-pyridineboronic acid pinacol ester ≥98.0% (by GC)
2-Chloro-4-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BClNO₂
MW: 239.51 g/mol
MDL Number: MFCD04039870
CAS Number: 458532-84-8

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 63.0 to 67.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR