4-Chlorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(4-Chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Chlorophenylboronic acid Pinacol ester, 4-Chlorobenzeneboronic acid pinacol ester, 1-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

C3500-25G C3500-5G
TCC3500-25GEA 147 USD
TCC3500-25G TCC3500-5G
4-Chlorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-Chlorophenylboronic acid pinacol ester
Formula: C₁₂H₁₆BClO₂
MDL Number: MFCD05663875
CAS Number: 195062-61-4

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 53.0 to 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR