2-Chlorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Chlorophenylboronic acid Pinacol ester, 2-(2-Chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

C3612-1G C3612-5G
TCC3612-1GEA 21 USD
TCC3612-1G TCC3612-5G
2-Chlorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Chlorophenylboronic acid pinacol ester
Formula: C₁₂H₁₆BClO₂
MDL Number: MFCD05663846
CAS Number: 870195-94-1

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) max. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR