2-Chloro-6-fluorobenzothiazole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Chloro-6-fluorobenzo[d]thiazole

C2607-1G C2607-5G
TCC2607-1GEA 101 USD
TCC2607-1G TCC2607-5G
2-Chloro-6-fluorobenzothiazole ≥98.0% (by GC)
2-Chloro-6-fluorobenzothiazole
Formula: C₇H₃ClFNS
MW: 187.62 g/mol
MDL Number: MFCD04448822
CAS Number: 399-74-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 91.0 to 95.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR