4-Bromooxindole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Bromooxindole, 4-Bromo-2,3-dihydro-1H-indol-2-one, 4-Bromoindolin-2-one

B5903-1G B5903-200MG
TCB5903-1GEA 187 USD
TCB5903-1G TCB5903-200MG
4-Bromooxindole ≥98.0% (by GC)
4-Bromooxindole
Formula: C₈H₆BrNO
MW: 212.05 g/mol
MDL Number: MFCD06659911
CAS Number: 99365-48-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 220.0 to 224.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR