3-Bromo-5-nitrobenzaldehyde ≥96.0% (by GC)

Supplier: TCI America
Warning

B4982-1G B4982-5G
TCB4982-1GEA 74 USD
TCB4982-1G TCB4982-5G
3-Bromo-5-nitrobenzaldehyde ≥96.0% (by GC)
3-Bromo-5-nitrobenzaldehyde
Formula: C₇H₄BrNO₃
MDL Number: MFCD00100097
CAS Number: 355134-13-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 100.0 to 104.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR