2-Bromo-3-iodopyridine ≥95.0% (by GC)

Supplier: TCI America
Warning

B5201-1G B5201-5G
TCB5201-1GEA 69 USD
TCB5201-1G TCB5201-5G
2-Bromo-3-iodopyridine ≥95.0% (by GC)
2-Bromo-3-iodopyridine
Formula: C₅H₃BrIN
MDL Number: MFCD09056782
CAS Number: 265981-13-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 96.0 to 100.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR