4-Bromobenzaldehyde dimethyl acetal ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-Bromo-4-(dimethoxymethyl)benzene, 4-Bromo-α,α-dimethoxytoluene

B5819-25G B5819-5G
TCB5819-25GEA 107 USD
TCB5819-25G TCB5819-5G
4-Bromobenzaldehyde dimethyl acetal ≥98.0% (by GC)
4-Bromobenzaldehyde dimethyl acetal
Formula: C₉H₁₁BrO₂
Boiling Pt: 94 °C
MDL Number: MFCD01320499
CAS Number: 24856-58-4

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR