4,5-Difluoro-2-methoxybenzonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

D5157-1G D5157-5G
TCD5157-1GEA 131 USD
TCD5157-1G TCD5157-5G
4,5-Difluoro-2-methoxybenzonitrile ≥98.0% (by GC)
4,5-Difluoro-2-methoxybenzonitrile
Formula: C₈H₅F₂NO
MDL Number: MFCD06660346
CAS Number: 425702-28-9
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 97.0 to 101.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR