2-Cyano-6-methylpyridine ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 6-Methylpicolinonitrile, 6-Methyl-2-pyridinecarbonitrile, 6-Cyano-2-picoline, 6-Methylpyridine-2-carbonitrile

C3025-1G C3025-5G
TCC3025-1GEA 44 USD
TCC3025-1G TCC3025-5G
2-Cyano-6-methylpyridine ≥98.0% (by GC)
2-Cyano-6-methylpyridine
Formula: C₇H₆N₂
MW: 118.14
Boiling Pt: 120 °C
MDL Number: MFCD00190586
CAS Number: 1620-75-3
EINECS: 216-588-9
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 74.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR