3-Amino-5-chloropyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-Chloropyridin-3-amine

A2876-1G A2876-5G
TCA2876-1GEA 82 USD
TCA2876-1G TCA2876-5G
3-Amino-5-chloropyridine ≥98.0% (by GC, titration analysis)
3-Amino-5-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
MDL Number: MFCD03701386
CAS Number: 22353-34-0

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 78.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR