2-Chloro-5-iodotoluene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-Chloro-4-iodo-2-methylbenzene

C3318-25G C3318-5G
TCC3318-25GEA 212 USD
TCC3318-25G TCC3318-5G
2-Chloro-5-iodotoluene ≥98.0% (by GC)
2-Chloro-5-iodotoluene
Formula: C₇H₆ClI
MW: 252.48 g/mol
Boiling Pt: 240 °C
Melting Pt: 10 °C
MDL Number: MFCD03093903
CAS Number: 116632-41-8

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR