3-Bromobenzenesulfonamide ≥98.0% (by GC)

Supplier: TCI America
Warning

B5734-25G B5734-5G
TCB5734-25GEA 124 USD
TCB5734-25G TCB5734-5G
3-Bromobenzenesulfonamide ≥98.0% (by GC)
3-Bromobenzenesulphonamide
Formula: C₆H₆BrNO₂S
MDL Number: MFCD00084903
CAS Number: 89599-01-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 156.0 to 160.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR