1-Bromo-3,5-dinitrobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3,5-Dinitrobromobenzene

B5627-1G B5627-5G
TCB5627-1GEA 35 USD
TCB5627-1G TCB5627-5G
1-Bromo-3,5-dinitrobenzene ≥98.0% (by GC)
1-Bromo-3,5-dinitrobenzene
Formula: C₆H₃BrN₂O₄
MW: 247 g/mol
MDL Number: MFCD00156596
CAS Number: 18242-39-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR