3-Bromo-5-fluorobenzonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

B4600-1G B4600-5G
TCB4600-1GEA 21 USD
TCB4600-1G TCB4600-5G
3-Bromo-5-fluorobenzonitrile ≥98.0% (by GC)
3-Bromo-5-fluorobenzonitrile
Formula: C₇H₃BrFN
MDL Number: MFCD04038227
CAS Number: 179898-34-1
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR