Di-tert-butylcarbodiimide ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1,3-Di-tert-butylcarbodiimide, N,N'-Di-tert-butylcarbodiimide, N,N-di-tert-butylcarbodiimide, N,N'-methanetetraylbis(1,1-dimethylethylamine)

D5347-25ML D5347-5ML
TCD5347-25MLEA 169 USD
TCD5347-25ML TCD5347-5ML
Di-tert-butylcarbodiimide ≥98.0% (by GC)
Di-tert-butylcarbodiimide
Formula: C₉H₁₈N₂
MW: 154.26 g/mol
Boiling Pt: 50 °C
Flash Pt: 35 °C
MDL Number: MFCD00010231
CAS Number: 691-24-7
EINECS: 211-719-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR