4-Amino-3-iodobenzonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

A2747-1G A2747-5G
TCA2747-1GEA 81 USD
TCA2747-1G TCA2747-5G
4-Amino-3-iodobenzonitrile ≥98.0% (by GC)
4-Amino-3-iodobenzonitrile
Formula: C₇H₅IN₂
MDL Number: MFCD04039964
CAS Number: 33348-34-4
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Amber powder to crystal
Purity (GC) min. 98.0 %
Melting point 113.0 to 117.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR