(S)-(-)-β-Citronellol ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: (S)-(-)-3,7-Dimethyl-6-octen-1-ol, (S)-(-)-ß-Citronellol, (S)-(-)-Citronellol, (-)-beta-Citronellol, (-)-β-Citronellol, (S)-3,7-Dimethyloct-6-en-1-ol, beta-Rhodinol

C3619-25ML C3619-5ML
TCC3619-25MLEA 85 USD
TCC3619-25ML TCC3619-5ML
(S)-(-)-β-Citronellol ≥98.0% (by GC)
(S)-(-)-β-Citronellol
Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 118 °C
Flash Pt: 102 °C
MDL Number: MFCD00063214
CAS Number: 7540-51-4
EINECS: 231-415-7

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 96.0 ee%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR