Bis(diethylamino)dimethylsilane ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: N,N,N',N'-Tetraethyl-1,1-dimethylsilanediamine

B5496-25ML B5496-5ML
TCB5496-25MLEA 110 USD
TCB5496-25ML TCB5496-5ML
Bis(diethylamino)dimethylsilane ≥98.0% (by GC)
Bis(diethylamino)dimethylsilane
Formula: C₁₀H₂₆N₂Si
Boiling Pt: 193 °C
Flash Pt: 35 °C
MDL Number: MFCD00039879
CAS Number: 4669-59-4
EINECS: 225-116-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR