5-Amino-3,4-dimethylisoxazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A3179-25G A3179-5G
TCA3179-25GEA 506 USD
TCA3179-25G TCA3179-5G
5-Amino-3,4-dimethylisoxazole ≥98.0% (by GC, titration analysis)
5-Amino-3,4-dimethylisoxazole
Formula: C₅H₈N₂O
MDL Number: MFCD00046081
CAS Number: 19947-75-2
EINECS: 243-437-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 119.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR