3-Chloroisonicotinonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

C2998-1G C2998-5G
TCC2998-1GEA 43 USD
TCC2998-1G TCC2998-5G
3-Chloroisonicotinonitrile ≥98.0% (by GC)
3-Chloroisonicotinonitrile
Formula: C₆H₃ClN₂
MDL Number: MFCD05663706
CAS Number: 68325-15-5
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR