5-Fluoropyridine-2-carbonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 5-Fluoro-2-pyridinecarbonitrile, 2-Cyano-5-fluoropyridine

C3286-1G C3286-5G
TCC3286-1GEA 26 USD
TCC3286-1G TCC3286-5G
5-Fluoropyridine-2-carbonitrile ≥98.0% (by GC)
5-Fluoropyridine-2-carbonitrile
Formula: C₆H₃FN₂
MW: 122.1 g/mol
MDL Number: MFCD08741371
CAS Number: 327056-62-2
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 40.0 to 44.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR