2-Bromoisonicotinonitrile ≥98.0% (by GC)
Supplier: TCI America
|
Danger
|
B5189-1G
B5189-5G
TCB5189-1GEA
38
USD
TCB5189-1G
TCB5189-5G
2-Bromoisonicotinonitrile ≥98.0% (by GC)
2-Bromoisonicotinonitrile
Formula:
C₆H₃BrN₂ MW: 183.01 g/mol |
MDL Number:
MFCD07367879 CAS Number: 10386-27-3 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...