3-Chloropropyl isocyanate ≥98.0% (by GC)

Supplier: TCI America
Danger

C3027-1G C3027-5G
TCC3027-1GEA 61 USD
TCC3027-1G TCC3027-5G
3-Chloropropyl isocyanate ≥98.0% (by GC)
3-Chloropropyl isocyanate
Formula: C₄H₆ClNO
Boiling Pt: 154 °C
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010384
CAS Number: 13010-19-0
UN: 2206
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR