4,4'-Dibromobenzophenone ≥98.0% (by GC)

Supplier: TCI America

D5211-1G D5211-5G
TCD5211-1GEA 38 USD
TCD5211-1G TCD5211-5G
4,4'-Dibromobenzophenone ≥98.0% (by GC)
4,4'-Dibromobenzophenone
Formula: C₁₃H₈Br₂O
Boiling Pt: 395 °C
MDL Number: MFCD00016330
CAS Number: 3988-03-2
EINECS: 223-632-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 174.0 to 178.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR