1,4-Bis(trimethylsilyl)benzene ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 1,4-Phenylenebis(trimethylsilane)

B2463-5G B2463-25G
TCB2463-5GEA 52 USD
TCB2463-5G TCB2463-25G
1,4-Bis(trimethylsilyl)benzene ≥97.0% (by GC)
1,4-Bis(trimethylsilyl)benzene
Formula: C₁₂H₂₂Si₂
MW: 222.48 g/mol
Boiling Pt: 194 °C
MDL Number: MFCD00015592
CAS Number: 13183-70-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 92.0 to 96.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR