1-(3-Chlorophenyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Danger

Synonyms: (3-Chlorophenyl)thiourea, 3-Chlorophenylthiourea, N-(3-Chlorophenyl)thiourea

C3168-1G C3168-5G
TCC3168-1GEA 43 USD
TCC3168-1G TCC3168-5G
1-(3-Chlorophenyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
1-(3-Chlorophenyl)-2-thiourea
Formula: (C₆H₄Cl)NC(S)NH₂
MW: 186.66 g/mol
MDL Number: MFCD00022165
CAS Number: 4947-89-1
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 137.0 to 141.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR