1-(2,6-Dimethylphenyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Danger

Synonyms: 1-(2,6-Dimethylphenyl)thiourea, (2,6-Dimethylphenyl)thiourea, 1-(2,6-Xylyl)thiourea, N-(2,6-Dimethylphenyl)thiourea, 2,6-Dimethylphenylthiourea

D4958-25G D4958-5G
TCD4958-25GEA 537 USD
TCD4958-25G TCD4958-5G
1-(2,6-Dimethylphenyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
1-(2,6-Dimethylphenyl)-2-thiourea
Formula: (C₆H₃(CH₃)₂)NC(S)NH₂
MW: 180.27 g/mol
MDL Number: MFCD00041165
CAS Number: 6396-76-5
EINECS: 229-005-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR