3-Amino-α-methylbenzyl alcohol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: m-Amino-α-methylbenzyl alcohol, 1-(3-Aminophenyl)ethanol, 3-Aminophenyl methyl carbinol, 3-(1-Hydroxyethyl)aniline

A3221-1G A3221-5G
TCA3221-1GEA 59 USD
TCA3221-1G TCA3221-5G
3-Amino-α-methylbenzyl alcohol ≥98.0% (by GC, titration analysis)
3-Amino-α-methylbenzyl alcohol
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 169 °C
MDL Number: MFCD00007798
CAS Number: 2454-37-7
EINECS: 219-525-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 67.0 to 71.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR