4'-Chloro-3'-fluoroacetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

C3298-1G C3298-5G
TCC3298-1GEA 50 USD
TCC3298-1G TCC3298-5G
4'-Chloro-3'-fluoroacetophenone ≥98.0% (by GC)
4'-Chloro-3'-fluoroacetophenone
Formula: C₈H₆ClFO
MDL Number: MFCD04115858
CAS Number: 151945-84-5

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR