3'-Chloro-5'-fluoroacetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-(3-Chloro-5-fluorophenyl)ethanone

C3288-1G C3288-5G
TCC3288-1GEA 44 USD
TCC3288-1G TCC3288-5G
3'-Chloro-5'-fluoroacetophenone ≥98.0% (by GC)
3'-Chloro-5'-fluoroacetophenone
Formula: C₈H₆ClFO
MW: 172.59 g/mol
MDL Number: MFCD04115834
CAS Number: 842140-52-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 55.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR