ω-Bromo-2-fluoroacetophenone ≥98.0% (by GC)

Supplier: TCI America
Danger

B5104-25G B5104-5G
TCB5104-25GEA 493 USD
TCB5104-25G TCB5104-5G
ω-Bromo-2-fluoroacetophenone ≥98.0% (by GC)
ω-Bromo-2-fluoroacetophenone
Formula: C₈H₆BrFO
Boiling Pt: 63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00278796
CAS Number: 655-15-2
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR