ω-Bromo-2,4-dimethoxyacetophenone ≥97.0% (by GC)

Danger

Synonyms: 2-Bromo-2',4'-dimethoxyacetophenone, 2,4-Dimethoxyphenacyl bromide, 2-Bromo-1-(2,4-dimethoxyphenyl)ethan-1-one, 2-Bromo-1-(2,4-dimethoxyphenyl)ethanone, α-Bromo-2,4-dimethoxyacetophenone

B4768-1G B4768-5G
TCB4768-1G TCB4768-5G
ω-Bromo-2,4-dimethoxyacetophenone ≥97.0% (by GC)
ω-Bromo-2,4-dimethoxyacetophenone
Formula: C₁₀H₁₁BrO₃
MW: 259.1 g/mol
MDL Number: MFCD00000197
CAS Number: 60965-26-6
EINECS: 262-542-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 103.0 to 107.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR