3-Chlorosalicylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

C3522-1G C3522-5G
TCC3522-1GEA 85 USD
TCC3522-1G TCC3522-5G
3-Chlorosalicylic acid ≥98.0% (by HPLC, titration analysis)
3-Chlorosalicylic acid
Formula: C₇H₅ClO₃
MW: 172.57 g/mol
MDL Number: MFCD00010390
CAS Number: 1829-32-9
EINECS: 217-379-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 182.0 to 186.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR