3,4-Dichlorobenzhydrazide ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3,4-Dichlorobenzenecarbohydrazide, 3,4-Dichlorobenzohydrazide, 3,4-Dichlorobenzene-1-carbohydrazide, 3,4-Dichlorobenzoic acid hydrazide, 3,4-Dichlorobenzoic hydrazide

D5392-1G D5392-5G
TCD5392-1GEA 68 USD
TCD5392-1G TCD5392-5G
3,4-Dichlorobenzhydrazide ≥98.0% (by GC)
3,4-Dichlorobenzhydrazide
Formula: C₇H₆Cl₂N₂O
MW: 205.04 g/mol
MDL Number: MFCD00017057
CAS Number: 28036-91-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 172.0 to 176.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR