4-Bromo-2-fluorobenzoyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 4-Bromo-2-fluorobenzene-1-carbonyl chloride

B5817-5G B5817-25G
TCB5817-5GEA 41 USD
TCB5817-5G TCB5817-25G
4-Bromo-2-fluorobenzoyl chloride ≥98.0% (by GC, titration analysis)
4-Bromo-2-fluorobenzoyl chloride
Formula: C₇H₃BrClFO
MDL Number: MFCD03411584
CAS Number: 151982-51-3
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR